missing translation for 'onlineSavingsMsg'
Learn More

2-Thiomorpholinoisonicotinaldehyde, 97%, Thermo Scientific™

Product Code. 10594764
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 898289-24-2
Molecular Formula C10H12N2OS
Molecular Weight (g/mol) 208.28
MDL Number MFCD09702361
InChI Key XESFZSHCANCGBY-UHFFFAOYSA-N
Synonym 2-thiomorpholinoisonicotinaldehyde, 2-thiomorpholin-4-yl pyridine-4-carbaldehyde, 2-thiomorpholin-4-yl pyridine-4-carboxaldehyde, 4-pyridinecarboxaldehyde,2-4-thiomorpholinyl, 2-thiomorpholin-4-yl isonicotinaldehyde, 2-1,4-thiazaperhydroin-4-yl pyridine-4-carbaldehyde
PubChem CID 24229507
IUPAC Name 2-thiomorpholin-4-ylpyridine-4-carbaldehyde
SMILES O=CC1=CC(=NC=C1)N1CCSCC1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 208.28
Percent Purity 97%
Chemical Name or Material 2-Thiomorpholinoisonicotinaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'