missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Thiohydantoin, 99%

Product Code. 11411447
Change view
Click to view available options
Quantity:
25g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11411447 25g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11411447 Supplier Alfa Aesar™ Supplier No. B24934.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 503-87-7
Molecular Formula C3H4N2OS
MDL Number MFCD00005277
Quantity 25g
Synonym 2-thiohydantoin, thiohydantoin, 4-imidazolidinone, 2-thioxo, 2-thioxoimidazolidin-4-one, 2-thioguidanthion, hydantoin, 2-thio, usaf be-25, 2-thioxo-4-imidazolidinone, 2-thioxo-1,3-diazolidin-4-one, glycine thiohydantoin
InChI Key UGWULZWUXSCWPX-UHFFFAOYSA-N
SMILES O=C1CNC(=S)N1
IUPAC Name 2-sulfanylideneimidazolidin-4-one
Molecular Weight (g/mol) 116.14
PubChem CID 1274030
Formula Weight 116.14
Percent Purity 99%
Chemical Name or Material 2-Thiohydantoin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.