missing translation for 'onlineSavingsMsg'
Learn More

2-Thienyl isocyanate, 90%, Maybridge

Product Code. 10632823
Change view
Click to view available options
:
Amber Glass Bottle; 1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10632823 Amber Glass Bottle; 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10632823 Supplier Thermo Scientific Maybridge Supplier No. CC13006DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 2048-57-9
CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber glass bottle
Molecular Formula C5H3NOS
MDL Number MFCD02677700
Quantity 1 g
Synonym 2-thienyl isocyanate, 2-thienylisocyanate, thiophene,2-isocyanato, thiophene, 2-isocyanato, thienyl isocyanate, isocyanatothiophene, thiophen-2-isocyanate, thien-2-yl isocyanate, 2-isocyanato-thiophene, 2-thiophene-yl isocyanate
InChI Key QVLWPBIUVXZGRK-UHFFFAOYSA-N
SMILES O=C=NC1=CC=CS1
IUPAC Name 2-isocyanatothiophene
Molecular Weight (g/mol) 125.15
PubChem CID 2776375
Formula Weight 125.15
Percent Purity 90%
Chemical Name or Material 2-Thienyl isocyanate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.