missing translation for 'onlineSavingsMsg'
Learn More

2-Thien-2-ylpyrimidine-5-carbaldehyde, 97%, Thermo Scientific™

Product Code. 10205464
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10205464 250 mg Amber Glass Bottle 250mg
10757485 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10205464 Supplier Thermo Scientific Maybridge Supplier No. CC66504CB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 921939-12-0
Molecular Formula C9H6N2OS
Molecular Weight (g/mol) 190.22
MDL Number MFCD09817553
InChI Key DCIBDJAGZUQNKM-UHFFFAOYSA-N
Synonym 2-thiophen-2-yl pyrimidine-5-carbaldehyde, 2-thien-2-ylpyrimidine-5-carbaldehyde, 2-thien-2-yl pyrimidine-5-carboxaldehyde, 2-2-thienyl pyrimidine-5-carbaldehyde, 2-thien-2-yl pyrimidine-5-carbaldehyde
PubChem CID 24229730
IUPAC Name 2-thiophen-2-ylpyrimidine-5-carbaldehyde
SMILES C1=CSC(=C1)C2=NC=C(C=N2)C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 190.23
Percent Purity 97%
Chemical Name or Material 2-Thien-2-ylpyrimidine-5-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.