missing translation for 'onlineSavingsMsg'
Learn More

2-Thien-2-ylbenzoic acid, 97%, Thermo Scientific™

Product Code. 10239752
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10239752 1 g Amber Glass Bottle 1g
10544103 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10239752 Supplier Thermo Scientific Maybridge Supplier No. CC35501DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 6072-49-7
Molecular Formula C11H8O2S
Molecular Weight (g/mol) 204.243
MDL Number MFCD04039152
InChI Key GDSOQCSYONDNAJ-UHFFFAOYSA-N
Synonym 2-2-thienyl benzoic acid, 2-thien-2-ylbenzoic acid, 2-thiophen-2-yl benzoic acid, thiolbenzoic acid, 2-2-thienyl-benzoic acid, 2-thiophen-2-yl-benzoic acid, 2-thiophene-2-yl-benzoic acid, benzoicacid, 2-2-thienyl, benzoic acid, 2-2-thienyl
PubChem CID 5105623
IUPAC Name 2-thiophen-2-ylbenzoic acid
SMILES C1=CC=C(C(=C1)C2=CC=CS2)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 204.25
Percent Purity 97%
Chemical Name or Material 2-Thien-2-ylbenzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.