missing translation for 'onlineSavingsMsg'
Learn More

2-Thenylmercaptan, 98%

Product Code. 10492821 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10492821 1 g Glass bottle 1g
10321614 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10492821 Supplier Thermo Scientific Acros Supplier No. 265110010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6258-63-5 | C5H6S2 | 130.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6258-63-5
Molecular Formula C5H6S2
Molecular Weight (g/mol) 130.22
MDL Number MFCD00107135
InChI Key GCZQHDFWKVMZOE-UHFFFAOYSA-N
Synonym 2-thiophenemethanethiol, thenyl mercaptan, 2-thenylmercaptan, 2-thienylmethanethiol, thiophene-2-methanethiol, 2-thenylthiol, thiophen-2-yl-methanethiol, unii-9jml279xs6, 2-thenyl mercaptan, 2-mercaptomethyl thiophene
PubChem CID 80408
IUPAC Name thiophen-2-ylmethanethiol
SMILES C1=CSC(=C1)CS

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.2000g/mL
Boiling Point 90°C (15.0 mmHg)
Flash Point 81°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.6020 to 1.604
Quantity 1 g
Specific Gravity 1.2
Solubility Information Solubility in water: slightly soluble
Formula Weight 130.22
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Thenylmercaptan
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.