missing translation for 'onlineSavingsMsg'
Learn More

2-(Tetrahydropyran-4-yloxy)benzonitrile, 97%, Thermo Scientific™

Product Code. 10757115
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 898289-37-7
Molecular Formula C12H13NO2
Molecular Weight (g/mol) 203.241
MDL Number MFCD09025873
InChI Key SXNKDLAJDADKEE-UHFFFAOYSA-N
Synonym 2-oxan-4-yloxy benzonitrile, 2-tetrahydropyran-4-yloxy benzonitrile, 2-oxan-4-yl oxy benzonitrile, 2-4-tetrahydropyranyl oxy benzonitrile, benzonitrile,2-tetrahydro-2h-pyran-4-yl oxy, 2-2h-3,4,5,6-tetrahydropyran-4-yloxy benzenecarbonitrile
PubChem CID 18525861
IUPAC Name 2-(oxan-4-yloxy)benzonitrile
SMILES C1COCCC1OC2=CC=CC=C2C#N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 203.24
Percent Purity 97%
Chemical Name or Material 2-(Tetrahydropyran-4-yloxy)benzonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'