missing translation for 'onlineSavingsMsg'
Learn More

2-(Tetrahydropyran-4-yloxy)benzonitrile, 97%, Thermo Scientific™

Product Code. 10432314
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10432314 1 g Amber Glass Bottle 1g
10757115 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10432314 Supplier Thermo Scientific Maybridge Supplier No. CC52216DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 898289-37-7
Molecular Formula C12H13NO2
Molecular Weight (g/mol) 203.241
MDL Number MFCD09025873
InChI Key SXNKDLAJDADKEE-UHFFFAOYSA-N
Synonym 2-oxan-4-yloxy benzonitrile, 2-tetrahydropyran-4-yloxy benzonitrile, 2-oxan-4-yl oxy benzonitrile, 2-4-tetrahydropyranyl oxy benzonitrile, benzonitrile,2-tetrahydro-2h-pyran-4-yl oxy, 2-2h-3,4,5,6-tetrahydropyran-4-yloxy benzenecarbonitrile
PubChem CID 18525861
IUPAC Name 2-(oxan-4-yloxy)benzonitrile
SMILES C1COCCC1OC2=CC=CC=C2C#N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 203.24
Percent Purity 97%
Chemical Name or Material 2-(Tetrahydropyran-4-yloxy)benzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.