missing translation for 'onlineSavingsMsg'
Learn More

2-Tetrahydrofuroic Acid, TRC

Product Code. 30425472
Change view
Click to view available options
Quantity:
5 g
50 g
Unit Size:
5g
Product Code. Quantity unitSize
30425472 5 g 5g
30417302 50 g -
2 options
This item is not returnable. View return policy

Product Code. 30425472

Brand: Toronto Research Chemicals TRCT2936455G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16874-33-2
Formula Weight 116.0473
InChI Formula InChI=1S/C5H8O3/c6-5(7)4-2-1-3-8-4/h4H,1-3H2,(H,6,7)
IUPAC Name oxolane-2-carboxylic acid
Molecular Formula C5H8O3
Molecular Weight (g/mol) 116.12
SMILES OC(=O)C1CCCO1
Synonym Tetrahydro-2-furancarboxylic Acid, Tetrahydro-2-furoic Acid, (R,S)-Tetrahydrofuran-2-carboxylic Acid, (±)-Tetrahydro-2-furoic Acid, (±)-Tetrahydrofuran-2-carboxylic Acid, 1,2,3,4-Tetrahydrofuran-2-carboxylic Acid, NSC 521564, Tetrahydro-2-furancarboxylic Acid
Chemical Name or Material 2-Tetrahydrofuroic Acid

Specifications

Quantity 5 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.