missing translation for 'onlineSavingsMsg'
Learn More

2-tert-Butylphenol, 99%

Product Code. 11347206
Change view
Click to view available options
Quantity:
100 g
500 g
2000 g
Unit Size:
100g
2kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11347206 500 g 500g
11337206 100 g 100g
11357206 2000 g 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11347206 Supplier Thermo Scientific Alfa Aesar Supplier No. L07294.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 88-18-6 | C10H14O | 150.221 g/mol

2-tert-Butylphenol is used pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-tert-Butylphenol is used pharmaceutical intermediates.

Solubility
Soluble in water (0.23 g/100 mL (20°C)).

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-18-6
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.221
MDL Number MFCD00002223
InChI Key WJQOZHYUIDYNHM-UHFFFAOYSA-N
Synonym 2-tert-butyl phenol, 2-t-butylphenol, phenol, 2-1,1-dimethylethyl, phenol, o-tert-butyl, o-t-butylphenol, o-tert-butylphenol, tert-butylphenol, 2-1,1-dimethylethyl phenol, 2-tert-butyl-1-hydroxybenzene, t-butylphenol
PubChem CID 6923
ChEBI CHEBI:34305
IUPAC Name 2-tert-butylphenol
SMILES CC(C)(C)C1=CC=CC=C1O

Specifications

Melting Point -7°C
Boiling Point 223°C to 224°C
Flash Point 102°C (215°F)
Quantity 500 g
UN Number UN2922
Beilstein 1907120
Formula Weight 150.22
Percent Purity 99%
Chemical Name or Material 2-tert-Butylphenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.