missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(tert-Butylamino)-3'-bromopropiophenone Hydrochloride, Mikromol™
Description
2-(tert-Butylamino)-3'-bromopropiophenone Hydrochloride
Specifications
Specifications
| Chemical Name or Material | 2-(tert-Butylamino)-3'-bromopropiophenone Hydrochloride |
| CAS | 1049718-43-5 |
| Analyte or Component Names | 2-(tert-Butylamino)-3'-bromopropiophenone Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C13 H18 Br N O . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | 2-(tert-Butylamino)-3'-bromopropiophenone Hydrochloride, GW 363083A, USP Bupropion Hydrochloride Related Compound B, 377U68, Bupropion Related Compound B as Hydrochloride |
| InChI Formula | InChI=1S/C13H18BrNO.ClH/c1-9(15-13(2,3)4)12(16)10-6-5-7-11(14)8-10;/h5-9,15H,1-4H3;1H |
| SMILES | Cl.CC(NC(C)(C)C)C(=O)c1cccc(Br)c1 |
| IUPAC Name | 1-(3-bromophenyl)-2-(tert-butylamino)propan-1-one;hydrochloride |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?