missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(tert-Butoxycarbonyloxyimino)phenylacetonitrile, TRC
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Chemical Name or Material | 2-(tert-Butoxycarbonyloxyimino)phenylacetonitrile |
| CAS | 58632-95-4 |
| Molecular Formula | C13H14N2O3 |
| Quantity | 25 g |
| Synonym | 2-(tert.-Butoxycarbonyloxyimino)phenylacetonitrile, Boc-ON, NSC 328381, alpha-[[[(1,1-Dimethylethoxy)carbonyl]oxy]imino]benzeneacetonitrile |
| InChI Formula | InChI=1S/C13H14N2O3/c1-13(2,3)17-12(16)18-15-11(9-14)10-7-5-4-6-8-10/h4-8H,1-3H3/b15-11+ |
| SMILES | CC(C)(C)OC(=O)O\N=C(/C#N)\c1ccccc1 |
| IUPAC Name | tert-butyl [(Z)-[cyano(phenyl)methylidene]amino] carbonate |
| Molecular Weight (g/mol) | 246.26 |
| Formula Weight | 246.1004 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction