missing translation for 'onlineSavingsMsg'
Learn More

2-Sulfobenzoic anhydride, 94%

Product Code. 11410913
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11410913 25 g 25g
11420913 100 g 100g
11430913 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11410913 Supplier Thermo Scientific Alfa Aesar Supplier No. A13314.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 81-08-3 | C7H4O4S | 184.165 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81-08-3
Molecular Formula C7H4O4S
Molecular Weight (g/mol) 184.165
MDL Number MFCD00005879
InChI Key NCYNKWQXFADUOZ-UHFFFAOYSA-N
Synonym 2-sulfobenzoic anhydride, 2-sulfobenzoic acid cyclic anhydride, sulfobenzoic anhydride, o-sulfobenzoic acid anhydride, 3h-2,1-benzoxathiol-3-one, 1,1-dioxide, 3h-benzo c 1,2 oxathiol-3-one 1,1-dioxide, unii-0swh68iq5w, o-sulfobenzoic acid, cyclic anhydride, 2,1-benzoxathiol-3-one-1,1-dioxide, 0swh68iq5w
PubChem CID 65729
IUPAC Name 1,1-dioxo-2,1$l^{6}-benzoxathiol-3-one
SMILES C1=CC=C2C(=C1)C(=O)OS2(=O)=O

Specifications

Melting Point ∼120°C
Boiling Point 184°C to 186°C (18 mmHg)
Quantity 25 g
Beilstein 139893
Sensitivity Moisture sensitive
Formula Weight 184.17
Percent Purity 94%
Chemical Name or Material 2-Sulfobenzoic anhydride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.