missing translation for 'onlineSavingsMsg'
Learn More

2-Sulfobenzoic acid hydrate, 98%

Product Code. p-7026978
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11480987 25 g 25g
11490987 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11480987 Supplier Thermo Scientific Alfa Aesar Supplier No. B23381.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 123333-68-6 | C7H6O5S | 202.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123333-68-6
Molecular Formula C7H6O5S
Molecular Weight (g/mol) 202.18
MDL Number MFCD00007479
InChI Key ZMPRRFPMMJQXPP-UHFFFAOYSA-N
Synonym 2-sulfobenzoic acid hydrate, benzoic acid, 2-sulfo-hydrate, acmc-20anmu, 2-sulphobenzoic acid hydrate, c7h6o5s.h2o, 2-sulfobenzoic acid, oxamethane, benzoic acid, 2-sulfo-, monohydrate
PubChem CID 24820382
IUPAC Name 2-sulfobenzoic acid;hydrate
SMILES OC(=O)C1=CC=CC=C1S(O)(=O)=O

Specifications

Melting Point ∼68°C to 72°C
Quantity 25 g
UN Number UN3261
Beilstein 2106852
Sensitivity Hygroscopic
Formula Weight 202.19 (Anhydrous)
Percent Purity 98%
Chemical Name or Material 2-Sulfobenzoic acid hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.