missing translation for 'onlineSavingsMsg'
Learn More

2-Sulfobenzoic acid cyclic anhydride, 95%

Product Code. 10675632 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10675632 100 g Glass bottle 100g
10541251 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10675632 Supplier Thermo Scientific Acros Supplier No. 184751000
 Restricted Item
Estimated Shipment: 19-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 81-08-3 | C7H4O4S | 184.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81-08-3
Molecular Formula C7H4O4S
Molecular Weight (g/mol) 184.17
MDL Number MFCD00005879
InChI Key NCYNKWQXFADUOZ-UHFFFAOYSA-N
Synonym 2-sulfobenzoic anhydride, 2-sulfobenzoic acid cyclic anhydride, sulfobenzoic anhydride, o-sulfobenzoic acid anhydride, 3h-2,1-benzoxathiol-3-one, 1,1-dioxide, 3h-benzo c 1,2 oxathiol-3-one 1,1-dioxide, unii-0swh68iq5w, o-sulfobenzoic acid, cyclic anhydride, 2,1-benzoxathiol-3-one-1,1-dioxide, 0swh68iq5w
PubChem CID 65729
IUPAC Name 1,1-dioxo-2,1$l^{6}-benzoxathiol-3-one
SMILES C1=CC=C2C(=C1)C(=O)OS2(=O)=O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 116°C to 127°C
Color White to Beige
Boiling Point 184°C to 186°C (18.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 100 g
Formula Weight 184.17
Percent Purity 95%
Grade Technical
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 2-Sulfobenzoic acid cyclic anhydride, tech., 90%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.