missing translation for 'onlineSavingsMsg'
Learn More

2-Sulfobenzaldehyde Sodium Salt, TRC

Product Code. 30427800
Change view
Click to view available options
Quantity:
50 g
100 g
250 g
Unit Size:
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30427800 250 g 250g
30414224 50 g -
30427801 100 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30427800 Supplier Toronto Research Chemicals Supplier No. TRCS689155250G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1008-72-6
Molecular Formula C7H5NaO4S
Molecular Weight (g/mol) 208.17
Synonym 2-Formylbenzenesulfonic Acid Sodium Salt, Benzaldehyde-2-sulfonic Acid Sodium Salt, Benzaldehyde-o-sulfonic Acid Sodium Salt, o-Formylbenzenesulfonic Acid Sodium Salt, o-Sulfobenzaldehyde Sodium Salt
IUPAC Name sodium;2-formylbenzenesulfonate
SMILES [Na+].[O-]S(=O)(=O)c1ccccc1C=O

Specifications

Quantity 250 g
InChI Formula InChI=1S/C7H6O4S.Na/c8-5-6-3-1-2-4-7(6)12(9,10)11;/h1-5H,(H,9,10,11);/q;+1/p-1
Formula Weight 207.9806
Chemical Name or Material 2-Sulfobenzaldehyde Sodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.