missing translation for 'onlineSavingsMsg'
Learn More

2-(Styrylthio)acetic acid, 97%, Thermo Scientific™

Product Code. 10405142
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10405142 10 g 10g
10580543 1 g 1g
10535934 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10405142 Supplier Thermo Scientific Maybridge Supplier No. S11590EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 13435-97-7
Molecular Formula C10H10O2S
Molecular Weight (g/mol) 194.248
MDL Number MFCD00068078
InChI Key IUEVBKDVPSQVLM-VOTSOKGWSA-N
Synonym 2-styrylthio acetic acid, e-2-phenylethenyl sulfanyl acetic acid, 2-phenylethenyl sulfanyl acetic acid, acetic acid,2-2-phenylethenyl thio, e-styrylsulfanyl-acetic acid, 2-1e-2-phenylvinylthio acetic acid, 2-e-2-phenylethenyl sulfanylacetic acid
PubChem CID 5474136
IUPAC Name 2-[(E)-2-phenylethenyl]sulfanylacetic acid
SMILES C1=CC=C(C=C1)C=CSCC(=O)O

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber glass bottle
Linear Formula C6H5CH=CHSCH2CO2H
Quantity 10 g
Formula Weight 194.25
Percent Purity 95%
Chemical Name or Material 2-(Styrylthio)acetic acid
compliance-icons
Product Identifier
  • 2-(Styrylthio)acetic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.