missing translation for 'onlineSavingsMsg'
Learn More

2-S-Glutathionyl Dimethyl Succinate (Mixture of Diastereomers), TRC

Product Code. 30394400
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30394400 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30394400 Supplier Toronto Research Chemicals Supplier No. TRCG3999501G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-S-Glutathionyl Dimethyl Succinate (Mixture of Diastereomers)
CAS 288847-14-3
Molecular Formula C16 H25 N3 O10 S
Quantity 1 g
Synonym L-γ-Glutamyl-S-[3-methoxy-1-(methoxycarbonyl)-3-oxopropyl]-L-cysteinylglycine, N5-((2R)-1-((Carboxymethyl)amino)-3-((1,4-dimethoxy-1,4-dioxobutan-2-yl)thio)-1-oxopropan-2-yl)-L-glutamine
InChI Formula InChI=1S/C16H25N3O10S/c1-28-13(23)5-10(16(27)29-2)30-7-9(14(24)18-6-12(21)22)19-11(20)4-3-8(17)15(25)26/h8-10H,3-7,17H2,1-2H3,(H,18,24)(H,19,20)(H,21,22)(H,25,26)/t8-,9-,10?/m0/s1
SMILES COC(=O)CC(SC[C@H](NC(=O)CC[C@H](N)C(=O)O)C(=O)NCC(=O)O)C(=O)OC
IUPAC Name (2S)-2-amino-5-[[(2R)-1-(carboxymethylamino)-3-(1,4-dimethoxy-1,4-dioxobutan-2-yl)sulfanyl-1-oxopropan-2-yl]amino]-5-oxopentanoic acid
Molecular Weight (g/mol) 451.449
Formula Weight 451.126
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.