missing translation for 'onlineSavingsMsg'
Learn More

2-Quinoxaloyl chloride, 95%

Product Code. 11936611 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11936611

Brand: Thermo Scientific Acros 163760010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 54745-92-5 | C9H5ClN2O | 192.6 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54745-92-5
Molecular Formula C9H5ClN2O
Molecular Weight (g/mol) 192.6
InChI Key SOPDQKNXOCUBSR-UHFFFAOYSA-N
Synonym 2-quinoxalinecarbonyl chloride, 2-quinoxalinecarbonylchloride, 2-quinoxaloyl chloride, 2-quinoxaloylchloride, 2-quinoxalinylcarbonyl chloride, 2-quinoxaline carbonyl chloride, 2-quinoxalinecarbonyl chloride 7ci, 9ci
PubChem CID 2734681
IUPAC Name quinoxaline-2-carbonyl chloride
SMILES C1=CC=C2C(=C1)N=CC(=N2)C(=O)Cl

Specifications

Melting Point 113.0°C to 115.0°C
Color Colorless to Yellow
CAS Min % 94.0
CAS Max % 100.0
Physical Form Liquid
Quantity 1 g
Assay Percent Range 94% min. (GC)
Formula Weight 192.6
Percent Purity 95%
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material 2-Quinoxaloyl chloride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Quinoxaloyl chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
  • EUH014-Reacts violently with water.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.