missing translation for 'onlineSavingsMsg'
Learn More

(2-Quinolyl)methylamine hydrochloride, 97%, Thermo Scientific™

Product Code. 10287803
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10287803 1 g Amber Glass Bottle 1g
10471494 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10287803 Supplier Thermo Scientific Maybridge Supplier No. CC04463DA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 861036-67-1
Molecular Formula C10H11ClN2
Molecular Weight (g/mol) 194.662
MDL Number MFCD09966142
InChI Key SUSYYDXUTFWSRV-UHFFFAOYSA-N
Synonym quinolin-2-ylmethanamine hydrochloride, 2-quinolyl methylamine hydrochloride, quinolin-2-yl methanamine hydrochloride, 2-aminomethylquinoline hydrochloride, 1-quinolin-2-yl methanamine hydrochloride, 2-quinolylmethylamine, chloride, 2-aminomethyl quinoline hydrochloride, quinolin-2-yl methylamine hydrochloride, quinolin-2-ylmethyl amine hydrochloride, c-quinolin-2-yl-methylamine hydrochloride
PubChem CID 18507529
IUPAC Name quinolin-2-ylmethanamine;hydrochloride
SMILES C1=CC=C2C(=C1)C=CC(=N2)CN.Cl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 194.66
Percent Purity 97%
Chemical Name or Material (2-Quinolyl)methylamine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.