missing translation for 'onlineSavingsMsg'
Learn More

2-Quinolinecarboxaldehyde, 97%

Product Code. 11926501 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11926501 25 g Glass bottle 25g
10324652 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11926501 Supplier Thermo Scientific Acros Supplier No. 370170250
 Restricted Item
Estimated Shipment: 11-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5470-96-2 | C10H7NO | 157.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5470-96-2
Molecular Formula C10H7NO
Molecular Weight (g/mol) 157.17
MDL Number MFCD00075032
InChI Key WPYJKGWLDJECQD-UHFFFAOYSA-N
Synonym 2-quinolinecarboxaldehyde, quinoline-2-carboxaldehyde, 2-quinolinecarbaldehyde, 2-formylquinoline, quinaldaldehyde, quinolinecarboxaldehyde, quinoline aldehyde, quinoline-2-aldehyde, zlchem 599, pubchem2277
PubChem CID 79619
IUPAC Name quinoline-2-carbaldehyde
SMILES O=CC1=CC=C2C=CC=CC2=N1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 66.0°C to 71.0°C
Color Orange to Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 25 g
Formula Weight 157.17
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 2-Quinolinecarboxaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • 2-Quinolinecarboxaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.