missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2-Pyrrolidone-5-carboxylic acid, sodium salt, 50 wt% solution in water

Product Code. 10491461 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10491461 25 mL 25mL
10023010 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10491461 Supplier Thermo Scientific Chemicals Supplier No. 232920250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 54571-67-4 | C5H6NNaO3 | 151.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Pyrrolidone-5-carboxylic acid, sodium salt
Color Colorless
Name Note 50 wt. % Solution in Water
CAS 54571-67-4, 7732-18-5
CAS Min % 47.5
CAS Max % 52.5
Molecular Formula C5H6NNaO3
MDL Number MFCD00066984
Density 1.27g/mL
Synonym sodium 5-oxopyrrolidine-2-carboxylate, sodium 5-oxo-dl-prolinate, monosodium 5-oxoproline, sodium 2-pyrrolidone-5-carboxylate, pca soda, proline, 5-oxo-, sodium salt 1:1, 2-pyrrolidone-5-carboxylic acid sodium salt, nalidone, 5-oxo-dl-proline, monosodium salt, dl-proline, 5-oxo-, monosodium salt
InChI Key CRPCXAMJWCDHFM-DFWYDOINSA-M
SMILES [Na+].[O-]C(=O)[C@@H]1CCC(=O)N1
IUPAC Name sodium;5-oxopyrrolidine-2-carboxylate
Molecular Weight (g/mol) 151.10
PubChem CID 23666346
Formula Weight 151.1
Percent Purity 47.5 to 52.5%
Packaging Glass bottle
pH 6.8 to 7.4
Quantity 25 mL
Physical Form Solution
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.