missing translation for 'onlineSavingsMsg'
Learn More

2-Pyrrolidinone, TRC

Product Code. 17999729
Change view
Click to view available options
Quantity:
25 g
50 g
100 g
Unit Size:
100g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17999729 25 g 25g
18031240 100 g 100g
18061330 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17999729 Supplier Toronto Research Chemicals Supplier No. TRCP99899025G
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 616-45-5
Molecular Formula C4 H7 N O
Molecular Weight (g/mol) 85.1
Synonym 2-Oxo-4-butyrolactam, 2-Oxopyrrolidine, 2-Pyrol, 2-Pyrrolidone, 2-Tetrahydropyrrolone, 4-Aminobutyric acid lactam, Azacyclopentan-2-one, Butanoic acid, 4-amino-, lactam, Butyrolactam, Kollisolv PYR, NSC 4593, NSC 8413, Pyrrolidin-2-one, Pyrrolidone, Soluphor P, α-Pyrrolidinone, α-Pyrrolidone, γ-Aminobutyric acid lactam, γ-Aminobutyric lactam, γ-Aminobutyrolactam, γ-Butyrolactam
IUPAC Name pyrrolidin-2-one
SMILES O=C1CCCN1

Specifications

Quantity 25 g
InChI Formula InChI=1S/C4H7NO/c6-4-2-1-3-5-4/h1-3H2,(H,5,6)
Formula Weight 85.0528
Percent Purity >95%
Chemical Name or Material 2-Pyrrolidone
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.