missing translation for 'onlineSavingsMsg'
Learn More

(2-Pyrrolidin-1-ylphenyl)methanol, 95%, Thermo Scientific™

Product Code. 10158314
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10158314 5 g Amber Glass Bottle 5g
10567494 250 mg Amber Glass Bottle 250mg
10337124 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10158314 Supplier Thermo Scientific Maybridge Supplier No. CC44809DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 73051-88-4
Molecular Formula C11H15NO
Molecular Weight (g/mol) 177.247
InChI Key QXIYEQQTAPJDBX-UHFFFAOYSA-N
Synonym 2-pyrrolidin-1-ylphenyl methanol, 2-pyrrolidin-1-yl phenyl methanol, 2-pyrrolizinobenzenemethanol, 2-1-pyrrolidinyl benzylalkohol, 2-pyrrolidinylphenyl methan-1-ol, benzenemethanol,2-1-pyrrolidinyl
PubChem CID 13021765
IUPAC Name (2-pyrrolidin-1-ylphenyl)methanol
SMILES C1CCN(C1)C2=CC=CC=C2CO

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 177.25
Percent Purity 95%
Chemical Name or Material (2-Pyrrolidin-1-ylphenyl)methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.