missing translation for 'onlineSavingsMsg'
Learn More

2-Pyrrolidin-1-ylisonicotinonitrile, ≥97%, Thermo Scientific™

Product Code. 10411514
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10411514 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10411514 Supplier Thermo Scientific Maybridge Supplier No. CC50416DA
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 127680-87-9
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C10H11N3
MDL Number MFCD08435908
Quantity 1 g
Synonym 2-pyrrolidin-1-yl isonicotinonitrile, 2-pyrrolidin-1-ylisonicotinonitrile, 2-pyrrolidin-1-yl pyridine-4-carbonitrile, 4-pyridinecarbonitrile,2-1-pyrrolidinyl, acmc-1c0ry, 2-pyrrolidinoisonicotinonitrile, 2-pyrrolidinylpyridine-4-carbonitrile
InChI Key POKSKCURNRHWPZ-UHFFFAOYSA-N
SMILES C1CCN(C1)C2=NC=CC(=C2)C#N
IUPAC Name 2-pyrrolidin-1-ylpyridine-4-carbonitrile
Molecular Weight (g/mol) 173.219
PubChem CID 14640855
Formula Weight 173.22
Percent Purity 97%
Chemical Name or Material 2-Pyrrolidin-1-ylisonicotinonitrile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.