missing translation for 'onlineSavingsMsg'
Learn More

2-Pyrrolidin-1-ylisonicotinic acid, ≥90%, Thermo Scientific™

Product Code. 11350677
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11350677 250 mg Amber Glass Bottle 250mg
10670454 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11350677 Supplier Thermo Scientific Maybridge Supplier No. CC50401CB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 98088-04-1
Molecular Formula C10H12N2O2
Molecular Weight (g/mol) 192.22
MDL Number MFCD04115212
InChI Key PQLHQGBNMJTAEH-UHFFFAOYSA-N
Synonym 2-pyrrolidin-1-yl-isonicotinic acid, 2-pyrrolidin-1-yl isonicotinic acid, 2-pyrrolidin-1-yl pyridine-4-carboxylic acid, 4-pyridinecarboxylic acid, 2-1-pyrrolidinyl, 2-pyrrolidin-1-ylisonicotinic acid, 2-1-pyrrolidinyl isonicotinic acid, 2-1-pyrrolidinyl-4-pyridinecarboxylic acid, 2-pyrrolidin-1-ylisonicotinic acid, 1.5 hydrate, 2-pyrrolidinylpyridine-4-carboxylic acid, hydrochloride
PubChem CID 6484236
IUPAC Name 2-pyrrolidin-1-ylpyridine-4-carboxylic acid
SMILES OC(=O)C1=CC(=NC=C1)N1CCCC1

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 219.25
Percent Purity 90%
Chemical Name or Material 2-Pyrrolidin-1-ylisonicotinic acid, 1.5 hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.