Learn More
2-Pyrrolidin-1-yl-1,3-thiazole-5-carboxylic acid, 97%, Thermo Scientific™
£34.55 - £85.15
Chemical Identifiers
CAS | 941716-91-2 |
---|---|
Molecular Formula | C8H10N2O2S |
Molecular Weight (g/mol) | 198.24 |
MDL Number | MFCD09879943 |
InChI Key | KPQGRYOIUITVHX-UHFFFAOYSA-N |
Synonym | 2-pyrrolidin-1-yl thiazole-5-carboxylic acid, 2-pyrrolidin-1-yl-1,3-thiazole-5-carboxylic acid, 2-pyrrolidinyl-1,3-thiazole-5-carboxylic acid |
PubChem CID | 24229715 |
SMILES | OC(=O)C1=CN=C(S1)N1CCCC1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10440944
|
Thermo Scientific Maybridge
CC64201CB |
250 mg | Amber Glass Bottle |
£34.55
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10644005
|
Thermo Scientific Maybridge
CC64201DA |
1 g | Amber Glass Bottle |
£85.15
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
941716-91-2 | |
198.24 | |
KPQGRYOIUITVHX-UHFFFAOYSA-N | |
24229715 |
C8H10N2O2S | |
MFCD09879943 | |
2-pyrrolidin-1-yl thiazole-5-carboxylic acid, 2-pyrrolidin-1-yl-1,3-thiazole-5-carboxylic acid, 2-pyrrolidinyl-1,3-thiazole-5-carboxylic acid | |
OC(=O)C1=CN=C(S1)N1CCCC1 |
Specifications
941716-91-2 | |
100.0 | |
C8H10N2O2S | |
2-pyrrolidin-1-yl thiazole-5-carboxylic acid, 2-pyrrolidin-1-yl-1,3-thiazole-5-carboxylic acid, 2-pyrrolidinyl-1,3-thiazole-5-carboxylic acid | |
OC(=O)C1=CN=C(S1)N1CCCC1 | |
198.24 | |
198.25 | |
2-Pyrrolidin-1-yl-1, 3-thiazole-5-carboxylic acid |
96.5 | |
97% | |
MFCD09879943 | |
KPQGRYOIUITVHX-UHFFFAOYSA-N | |
2-(pyrrolidin-1-yl)-1,3-thiazole-5-carboxylic acid | |
24229715 | |
97% |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if inhaled.
Harmful in contact with skin.
Causes skin irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning