missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Pyrimidinecarbonitrile, 98%, Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
25 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
CAS Min % | 97.5 |
CAS Max % | 100.0 |
Melting Point | 37.0°C to 41.0°C |
Color | White to Yellow |
Flash Point | 110°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (GC) |
Packaging | Glass bottle |
Quantity | 25 g |
Solubility Information | Solubility in water: .. Other solubilities: soluble in methanol |
Show More |
Safety and Handling
Product Identifier
- 2-Pyrimidinecarbonitrile
- Danger
- Acute toxicity Category 3
- Respiratory sensitiser Category 1
- H317-May cause an allergic skin reaction.
- H301-Toxic if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'