missing translation for 'onlineSavingsMsg'
Learn More

2-Pyridyl trifluoromethanesulfonate, 98%, Thermo Scientific™

Product Code. 15452228
Change view
Click to view available options
Quantity:
5 mL
25 mL
Unit Size:
25mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15452228 25 mL 25mL
15442228 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15452228 Supplier Thermo Scientific Alfa Aesar Supplier No. H55855.AC

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65007-00-3
Molecular Formula C6H4F3NO3S
Molecular Weight (g/mol) 227.157
MDL Number MFCD00192531
InChI Key COLRMVLTWJTLFJ-UHFFFAOYSA-N
Synonym 2-pyridyl trifluoromethanesulfonate, 2-pyridyl triflate, pyridyl triflate, acmc-1bdfn, 2-pyridyltrifluoromethanesulfonate, 2-pyridyl trifluoromethane-sulfonate, pyridin-2-yltrifluoromethanesulfonate, 2-trifluoro-methylsulfonyloxy-pyridine, trifluoromethanesulfonic acid 2-pyridyl ester
PubChem CID 3947626
IUPAC Name pyridin-2-yl trifluoromethanesulfonate
SMILES C1=CC=NC(=C1)OS(=O)(=O)C(F)(F)F

Specifications

Color Colorless to Orange
Density 1.477g/mL
Boiling Point 108°C (25 mmHg)
Refractive Index 1.4350
Quantity 25 mL
UN Number UN1993
Formula Weight 227.16
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Pyridyl trifluoromethanesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.