missing translation for 'onlineSavingsMsg'
Learn More

2-Pyridinesulfonic acid, 97%, Thermo Scientific Chemicals

Product Code. 15781749 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15781749 5 g 5g
15771749 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15781749 Supplier Thermo Scientific Acros Supplier No. 455760050
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 15103-48-7 | Molecular Formula: C5H5NO3S | Molecular Weight: 159.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15103-48-7
Molecular Formula C5H5NO3S
Molecular Weight (g/mol) 159.16
MDL Number MFCD00234080
InChI Key KZVLNAGYSAKYMG-UHFFFAOYSA-N
Synonym 2-pyridinesulfonic acid, pyridinesulfonic acid, 2-pyridinesulfonic acid 8ci 9ci, alpha-pyridinesulfonic acid, sulfopyridin, pyridine-2-sulfonicacid, acmc-1bo1h, .alpha.-pyridinesulfonic acid, kzvlnagysakymg-uhfffaoysa
PubChem CID 94849
IUPAC Name pyridine-2-sulfonic acid
SMILES OS(=O)(=O)C1=CC=CC=N1

Specifications

Melting Point 244.0°C to 249.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 5 g
Solubility Information Solubility in water: very soluble.
Formula Weight 159.16
Percent Purity 97%
Chemical Name or Material 2-Pyridinesulfonic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.