missing translation for 'onlineSavingsMsg'
Learn More

2-Pyridinemethanol, 98+%

Product Code. 11497227
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11497227

Brand: Thermo Scientific Alfa Aesar A15005.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 586-98-1 | C6H7NO | 109.128 g/mol

2-Pyridinemethanol is used as a pyridylalcohol with hypoglycemic activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Pyridinemethanol is used as a pyridylalcohol with hypoglycemic activity.

Solubility
Fully miscible in water.

Notes
Hygroscopic. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 586-98-1
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.128
MDL Number MFCD00006348
InChI Key SHNUBALDGXWUJI-UHFFFAOYSA-N
Synonym 2-pyridinemethanol, 2-hydroxymethyl pyridine, piconol, 2-pyridinylmethanol, 2-pyridylmethanol, 2-pyridylcarbinol, pyridine-2-methanol, pyridine-2-carbinol, alpha-picolyl alcohol, pyridin-2-yl methanol
PubChem CID 11474
IUPAC Name pyridin-2-ylmethanol
SMILES C1=CC=NC(=C1)CO

Specifications

Melting Point -6°C to -5°C
Density 1.131
Boiling Point 112°C to 113°C (16 mmHg)
Flash Point 120°C (248°F)
Refractive Index 1.544
Quantity 25 g
Beilstein 107849
Sensitivity Hygroscopic
Solubility Information Fully miscible in water.
Formula Weight 109.13
Percent Purity ≥98%
Chemical Name or Material 2-Pyridinemethanol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.