missing translation for 'onlineSavingsMsg'
Learn More

2-Pyridineethanol, TRC

Product Code. 30420813
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30420813 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30420813 Supplier Toronto Research Chemicals Supplier No. TRCP991520500G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(Pyridin-2-yl)ethanol (2-(2-Hydroxyethyl)pyridine)
CAS 103-74-2
Purity Grade Notes HPLC
Molecular Formula C7 H9 N O
Quantity 500 g
Synonym 2-(Pyridin-2-yl)ethanol, Betahistine Dihydrochloride Imp. B (EP), 2-(2-Hydroxyethyl)pyridine, 2-Pyridineethanol, 2-(2-Hydroxylethyl)pyridine, 2-(2-Pyridinyl)ethanol, 2-(2-Pyridyl)ethanol, 2-(2-Pyridyl)ethyl alcohol, 2-(α-Pyridyl)ethanol, 2-(β-Hydroxyethyl)pyridine, 2-Pyridylethanol, NSC 2144, NSC 77979, β-(2-Pyridyl)ethanol
InChI Formula InChI=1S/C7H9NO/c9-6-4-7-3-1-2-5-8-7/h1-3,5,9H,4,6H2
SMILES OCCc1ccccn1
IUPAC Name 2-pyridin-2-ylethanol
Molecular Weight (g/mol) 123.15
Formula Weight 123.0684
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.