missing translation for 'onlineSavingsMsg'
Learn More

2-Pyridinecarboxylic acid, 97%, Thermo Scientific™

Catalog Number p-2698790
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
98-98-6
C6H5NO2
123.111
MFCD00006293
SIOXPEMLGUPBBT-UHFFFAOYSA-N
picolinic acid, 2-pyridinecarboxylic acid, 2-picolinic acid, 2-carboxypyridine, alpha-picolinic acid, o-pyridinecarboxylic acid, pyridinecarboxylic acid, alpha-pyridinecarboxylic acid, picolinate, acide picolique
1018
CHEBI:28747
pyridine-2-carboxylic acid
C1=CC=NC(=C1)C(=O)O
This item is not returnable. View return policy
98-98-6
C6H5NO2
123.111
MFCD00006293
SIOXPEMLGUPBBT-UHFFFAOYSA-N
picolinic acid, 2-pyridinecarboxylic acid, 2-picolinic acid, 2-carboxypyridine, alpha-picolinic acid, o-pyridinecarboxylic acid, pyridinecarboxylic acid, alpha-pyridinecarboxylic acid, picolinate, acide picolique
1018
CHEBI:28747
pyridine-2-carboxylic acid
C1=CC=NC(=C1)C(=O)O
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 123.11
Percent Purity 97%
Chemical Name or Material 2-Pyridinecarboxylic acid
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'