missing translation for 'onlineSavingsMsg'
Learn More

2-Pyridinecarboxaldehyde, 99%

Product Code. 10434405 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10434405 500 g Glass bottle 500g
10755611 25 g Glass bottle 25g
10173420 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 10434405 missing translation for 'mfr' Thermo Scientific Acros missing translation for 'supplierNo' 131825000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1121-60-4 | C6H5NO | 107.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1121-60-4
Molecular Formula C6H5NO
Molecular Weight (g/mol) 107.11
MDL Number MFCD00006290
InChI Key CSDSSGBPEUDDEE-UHFFFAOYSA-N
Synonym 2-pyridinecarboxaldehyde, picolinaldehyde, pyridine-2-carboxaldehyde, 2-formylpyridine, pyridine-2-aldehyde, picolinal, 2-picolinaldehyde, 2-pyridaldehyde, picolinic aldehyde, 2-pyridylaldehyde
PubChem CID 14273
ChEBI CHEBI:73012
IUPAC Name pyridine-2-carbaldehyde
SMILES C1=CC=NC(=C1)C=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Brown-Yellow to Yellow
Density 1.1210g/mL
Boiling Point 180.0°C to 182.0°C (761.0 mmHg)
Flash Point 73°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (C5H4N)COH
Refractive Index 1.5343 to 1.5373
Quantity 500 g
Beilstein 21, I, 287
Specific Gravity 1.121
Solubility Information Solubility in water: miscible. Other solubilities: soluble in most common organic solvents
Formula Weight 107.11
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Pyridinecarboxaldehyde
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Pyridinecarboxaldehyde
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H330-Fatal if inhaled.
  • H411-Toxic to aquatic life with long lasting effects.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.