missing translation for 'onlineSavingsMsg'
Learn More

2-Pyridin-3-ylpiperidine, Thermo Scientific™

Product Code. 11376915
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11376915 5 g Amber Glass Bottle 5g
10599582 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11376915 Supplier Thermo Scientific Maybridge Supplier No. RJC03825DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 13078-04-1
Molecular Formula C10H14N2
Molecular Weight (g/mol) 162.24
MDL Number MFCD00006370
InChI Key MTXSIJUGVMTTMU-UHFFFAOYNA-N
Synonym 3-piperidin-2-yl pyridine, anabasine, dl-anabasine, neonicotine, +--anabasine, 2-pyridin-3-ylpiperidine, +/--anabasine, 3-2-piperidinyl pyridine, 2-3-pyridyl piperidine, pyridine, 3-2-piperidinyl
PubChem CID 2181
ChEBI CHEBI:28986
SMILES C1CCC(NC1)C1=CC=CN=C1

Specifications

Density 1.0460g/mL
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
IUPAC Name 3-(piperidin-2-yl)pyridine
Formula Weight 162.23
Percent Purity 97%
Chemical Name or Material 2-Pyridin-3-ylpiperidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.