missing translation for 'onlineSavingsMsg'
Learn More

2-Pyrid-4-ylpyrimidine-5-carboxylic acid, 90%, Thermo Scientific™

Product Code. 10022354
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10022354 1 g Amber Glass Bottle 1g
10346764 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10022354 Supplier Thermo Scientific Maybridge Supplier No. CC66601DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 259807-47-1
Molecular Formula C10H7N3O2
Molecular Weight (g/mol) 201.185
MDL Number MFCD09863218
InChI Key GTKJGOOMSODGOU-UHFFFAOYSA-N
Synonym 2-pyridin-4-yl pyrimidine-5-carboxylic acid, 2-pyrid-4-ylpyrimidine-5-carboxylic acid, 5-pyrimidinecarboxylicacid, 2-4-pyridinyl, 2-4-pyridyl pyrimidine-5-carboxylic acid, 2-4-pyridinyl-5-pyrimidinecarboxylic acid, 2-pyridin-4-yl-pyrimidine-5-carboxylic acid, 5-pyrimidinecarboxylic acid, 2-4-pyridinyl, 5-pyrimidinecarboxylic acid, 2-4-pyridinyl-9ci
PubChem CID 22710213
IUPAC Name 2-pyridin-4-ylpyrimidine-5-carboxylic acid
SMILES C1=CN=CC=C1C2=NC=C(C=N2)C(=O)O

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 201.18
Percent Purity 90%
Chemical Name or Material 2-Pyrid-4-ylpyrimidine-5-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.