missing translation for 'onlineSavingsMsg'
Learn More

2-Pyrazinylmethanol, 97%, Thermo Scientific™

Product Code. 10059081
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10059081 1 g Amber Glass Bottle 1g
10722663 250 mg Amber Glass Bottle 250mg
10526314 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10059081 Supplier Thermo Scientific Maybridge Supplier No. CC03409DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 6705-33-5
Molecular Formula C5H6N2O
Molecular Weight (g/mol) 110.12
MDL Number MFCD01693938
InChI Key LFCWHDGQCWJKCG-UHFFFAOYSA-N
Synonym 2-pyrazinylmethanol, pyrazin-2-yl methanol, pyrazinemethanol, 2-hydroxymethyl pyrazine, 2-pyrazinemethanol, 2-hydroxymethylpyrazine, pyrazin-2-yl-methanol, unii-c27z7qi77d, pyrazin-2-ylmethan-1-ol
PubChem CID 201734
SMILES OCC1=CN=CC=N1

Especificaciones

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
IUPAC Name pyrazin-2-ylmethanol
Formula Weight 110.12
Percent Purity 95%
Chemical Name or Material 2-Pyrazinylmethanol
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.