missing translation for 'onlineSavingsMsg'
Learn More

2-Propionylthiazole, 99%

Product Code. 11410317
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11410317 10 g 10g
11400317 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11410317 Supplier Thermo Scientific Alfa Aesar Supplier No. B21711.09
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 43039-98-1 | C6H7NOS | 141.188 g/mol

Used as flavoring agent and fragrance chemical compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as flavoring agent and fragrance chemical compounds.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 43039-98-1
Molecular Formula C6H7NOS
Molecular Weight (g/mol) 141.188
MDL Number MFCD01681404
InChI Key TYRAENAWSLPSLW-UHFFFAOYSA-N
Synonym 2-propionylthiazole, 1-thiazol-2-yl propan-1-one, 2-propionyl thiazole, thiazole, 2-propionyl, 1-propanone, 1-2-thiazolyl, 1-2-thiazolyl-1-propanone, ethyl 2-thiazolyl ketone, 1-1,3-thiazol-2-yl propan-1-one, 2-propionylthiazol, unii-05cb62uh9k
PubChem CID 65288
IUPAC Name 1-(1,3-thiazol-2-yl)propan-1-one
SMILES CCC(=O)C1=NC=CS1

Specifications

Density 1.174
Boiling Point 110°C (5 mmHg)
Flash Point >100°C (212°F)
Quantity 10 g
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 141.19
Percent Purity 99%
Chemical Name or Material 2-Propionylthiazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.