missing translation for 'onlineSavingsMsg'
Learn More

2-Propionylpyrrole, 99%

Product Code. 11496826
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11496826 5 g 5g
11486826 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Produktkod. 11496826 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer B23954.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

CAS: 1073-26-3 | C7H9NO | 123.155 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 1073-26-3
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.155
MDL Number MFCD01696449
InChI Key AEJPPSRYXGEVDT-UHFFFAOYSA-N
Synonym 2-propionylpyrrole, 1-1h-pyrrol-2-yl propan-1-one, 2-pyrryl ethyl ketone, ethyl 2-pyrrolyl ketone, alpha-propionylpyrrole, 1-2-pyrrolyl-1-propanone, alpha-pyrryl ethyl ketone, 1-propanone, 1-pyrrol-2-yl, 1-1h-pyrrol-2-yl-1-propanone, unii-hgs11xf57f
PubChem CID 61260
IUPAC Name 1-(1H-pyrrol-2-yl)propan-1-one
SMILES CCC(=O)C1=CC=CN1

Specifikationer

Melting Point 48°C to 51°C
Quantity 5 g
Formula Weight 123.15
Percent Purity 99%
Chemical Name or Material 2-Propionylpyrrole

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.