missing translation for 'onlineSavingsMsg'
Learn More

2-Propionylpyrrole, 99%

Product Code. 11486826
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11486826 1 g 1g
11496826 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11486826 Supplier Thermo Scientific Alfa Aesar Supplier No. B23954.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1073-26-3 | C7H9NO | 123.155 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1073-26-3
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.155
MDL Number MFCD01696449
InChI Key AEJPPSRYXGEVDT-UHFFFAOYSA-N
Synonym 2-propionylpyrrole, 1-1h-pyrrol-2-yl propan-1-one, 2-pyrryl ethyl ketone, ethyl 2-pyrrolyl ketone, alpha-propionylpyrrole, 1-2-pyrrolyl-1-propanone, alpha-pyrryl ethyl ketone, 1-propanone, 1-pyrrol-2-yl, 1-1h-pyrrol-2-yl-1-propanone, unii-hgs11xf57f
PubChem CID 61260
IUPAC Name 1-(1H-pyrrol-2-yl)propan-1-one
SMILES CCC(=O)C1=CC=CN1

Specifications

Melting Point 48°C to 51°C
Quantity 1 g
Formula Weight 123.15
Percent Purity 99%
Chemical Name or Material 2-Propionylpyrrole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.