missing translation for 'onlineSavingsMsg'
Learn More

2-Propionylbenzoic Acid, TRC

Product Code. 30427026
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30427026 100 mg 100mg
30416913 500 mg 500mg
30409326 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30427026 Supplier Toronto Research Chemicals Supplier No. TRCP839045100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2360-45-4
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.18
Synonym 2-(1-Oxopropyl)benzoic Acid, 2-(1-Oxopropyl)-1-benzoic Acid, 2-Propanoylbenzoic Acid, Propiophenone-2-carboxylic Acid
IUPAC Name 2-propanoylbenzoic acid
SMILES CCC(=O)c1ccccc1C(=O)O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C10H10O3/c1-2-9(11)7-5-3-4-6-8(7)10(12)13/h3-6H,2H2,1H3,(H,12,13)
Formula Weight 178.063
Chemical Name or Material 2-Propionylbenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.