missing translation for 'onlineSavingsMsg'
Learn More

2-Propene-1-sulfonic acid sodium salt, 95%, pract.

Product Code. 10249700 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10249700 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10249700 Supplier Thermo Scientific Acros Supplier No. 418370250
 Restricted Item
Estimated Shipment: 02-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2495-39-8 | C3H5NaO3S | 144.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 2495-39-8
CAS Min % 94.0
CAS Max % 100.0
Color White
Flash Point >100°C
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Molecular Formula C3H5NaO3S
Linear Formula CH2CHCH2SO3Na
MDL Number MFCD00051416
Quantity 25 g
Synonym sodium allylsulfonate, sodium prop-2-ene-1-sulfonate, 2-propene-1-sulfonic acid, sodium salt, sodium allyl sulfonate, sodium 2-propene-1-sulfonate, unii-7sgf7tb9o2, sodium prop-2-enesulphonate, allylsulfonic acid sodium salt, sodium allysulfonate, 2-propene-1-sulfonic acid sodium salt
Solubility Information Solubility in water: 1g/25mL. Other solubilities: soluble in alcohol
InChI Key DIKJULDDNQFCJG-UHFFFAOYSA-M
SMILES C=CCS(=O)(=O)[O-].[Na+]
IUPAC Name sodium;prop-2-ene-1-sulfonate
Molecular Weight (g/mol) 144.13
PubChem CID 23690996
Formula Weight 144.13
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 2-Propene-1-sulfonic acid sodium salt, pract., 95%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.