missing translation for 'onlineSavingsMsg'
Learn More

2-Propanol, Semiconductor Grade, 99.5% min

Product Code. 11363567
Change view
Click to view available options
Quantity:
500 mL
1 L
4 x 1 L
Unit Size:
1 L
4 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11363567 1 L 1 L
11420500 500 mL 500mL
11343567 4 x 1 L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11363567 Supplier Thermo Scientific Alfa Aesar Supplier No. 019397.K2
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Maximum level of impurities: Color (APHA) 10, Evaporation residue 5ppm, Solubility in H2O P.T., H2O (by Karl Fischer titrn) 0.05%, Acidity 0.2meq/g, Alkalinity 0.1meq/g, Cl 0.2ppm, PO4 0.5ppm | CAS: 67-63-0 | C3H8O | 60.096 g/mol

2-Propanol is used as a solvent for gums, resins, alkaloids and essential oils, as a denaturant for ethyl alcohol and is used in lotions and cosmetics. It is used in keyboard, LCD and laptop cleaning, as a wetting agent in the fountain solution used in lithographic printing, and often used as a solvent for French polishing shellac used in cabinet making. It acts as hydride source in the Meerwein-Ponndorf-Verley reduction. It plays an important role in DNA extraction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
MDL Number MFCD00011674
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Melting Point °C to 90°C
Color Colorless
Density 0.785
Boiling Point 82°C to 83°C
Flash Point 12°C (53°F)
Ignition Point 455°C (851°F)
Assay Percent Range 99.5% min.
Odor Alcohol-like
Refractive Index 1.377
Quantity 1 L
Beilstein 635639
Solubility Information Miscible with water,ethanol,acetone,chloroform,methanol,diethyl ether,n-octanol and benzene.
Formula Weight 60.1
Vapor Pressure 48 hPa (36mm Hg at 20°C)
Grade Semiconductor
Concentration or Composition (by Analyte or Components) 2-Propanol
Physical Form Liquid
Chemical Name or Material 2-Propanol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Propanol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 2-Propanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.