missing translation for 'onlineSavingsMsg'
Learn More

2-Propanol, Puriss. p.a., ACS Reagent, Reag. ACS, Reag. ISO, Reag. Ph. Eur., Reag. USP, Solstice™

Product Code. 16625923
Change view
Click to view available options
Quantity:
4 x 2.5 L
6 x 1 L
Unit Size:
2.50 L
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16625923 6 x 1 L Pack of 6
16615923 4 x 2.5 L 2.50 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16625923 Supplier Fluka Supplier No. 722996X1L
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Melting Point -90°C
Color Colorless
CAS Max % 1
Density 0.784 - 0.786 g/cm3 (20°C)
Quantity 6 x 1 L
Flash Point 12°C
Boiling Point 77°C to 83°C (1013 hPa)
Chemical Name or Material 2-Propanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.