missing translation for 'onlineSavingsMsg'
Learn More

2-Propanol, puriss., Honeywell
BIOPHARMA

Product Code. 15672520
Change view
Click to view available options
Quantity:
100 mL
1 L
2.5 L
5 L
10 L
20 kg
25 L
160 kg
Unit Size:
1 L
10 L
100mL
160kg
2.5 L
20kg
25 L
5 L
Product Code. Quantity unitSize
15672520 20 kg 20kg
15612520 100 mL 100mL
15652520 1 L 1 L
15662520 2.5 L 2.5 L
15612530 5 L 5 L
15622520 10 L 10 L
15682520 25 L 25 L
15642520 160 kg 160kg
8 options
This item is not returnable. View return policy

Product Code. 15672520

Brand: Honeywell Riedel-de Haën 2413720KG

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
MDL Number MFCD00011674
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Density 0.785g/mL at 25°C
Quantity 20 kg
Explosion Limit 0.02
Flash Point 12°C (53.6°F)
Ignition Point 750°F
Linear Formula (CH3)2CHOH
Vapor Pressure 33mmHg (20°C), 44mmHg (25°C)
UN Number 1219
Beilstein 635639
Formula Weight 60.1g/mol
Percent Purity ≥99.5% (GC)
Grade puriss.
Chemical Name or Material 2-Propanol, Meets analytical specification of Ph. Eur.BP, USP
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Propanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.