missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
2-Propanol, >-99.90%, C3H8O, CAS Number-67-63-0, petrohol, isopropyl alcohol, isopropanol, sec-propyl alcohol, dimethylcarbinol, 2-hydroxypropane, hartosol, 2-propanol, alkolave, avantine, 4L, Report AU at 254nm, Report AU at 210nm, Report AU at 230nm, Report AU at 220nm
Specifications
Specifications
| pH | 7 |
| Melting Point | -89.5°C |
| Absorbance | Report AU at 210nm, Report AU at 220nm, Report AU at 230nm, Report AU at 254nm |
| Color | Colorless |
| Density | 0.785g/cm³ |
| Quantity | 4 L |
| Identification | Pass Test |
| Packaging | Glass Bottle |
| Physical Form | Liquid |
| Residue after Evaporation | 1ppm max. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?