missing translation for 'onlineSavingsMsg'
Learn More

2-Propanol-d{8}, 99%(Isotopic), Thermo Scientific Chemicals

Catalog Number 11374368
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
22739-76-0
C3H8O
68.15
MFCD00044341
KFZMGEQAYNKOFK-PIODKIDGSA-N
2-propanol-d8, isopropanol-d8, 2 h? propan-2-2 h ol, octadeuteroisopropanol, 2h8-2-propanol, 2-propan-1,1,1,2,3,3,3-d7-ol-d, 2-propanol-d isotopic 1g, 2-propanol-d8, 99.5 atom % d
2723972
[2H]OC([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H]
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
22739-76-0
C3H8O
68.15
MFCD00044341
KFZMGEQAYNKOFK-PIODKIDGSA-N
2-propanol-d8, isopropanol-d8, 2 h? propan-2-2 h ol, octadeuteroisopropanol, 2h8-2-propanol, 2-propan-1,1,1,2,3,3,3-d7-ol-d, 2-propanol-d isotopic 1g, 2-propanol-d8, 99.5 atom % d
2723972
[2H]OC([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H]

2-Propanol-d{8} is used as an intermediate in chemical research and in pharmaceutics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Liquid
Density 0.90 g/mL
Flash Point −11.7°C (53°F)
Odor Alcohol-like
Quantity 5 g
Melting Point -89.5°C
Boiling Point 82.4°C
UN Number UN1219
Solubility Information Completely soluble in water.
IUPAC Name (²H₇)propan-2-(²H)ol
Formula Weight 68.15
Percent Purity 99%
Chemical Name or Material 2-Propanol-d8, Isotopic
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'