missing translation for 'onlineSavingsMsg'
Learn More

2-Propanol, anhydrous, 99.5+%, packaged under Argon in resealable ChemSeal™ bottles

Product Code. 11354807
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
This item is not returnable. View return policy

Product Code. 11354807

Brand: Thermo Scientific Alfa Aesar 041840.AK

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 67-63-0 | C3H8O | 60.096 g/mol

2-Propanol acts as a solvent for pharmaceuticals, perfumes, gums, resins, alkaloids and essential oils. It is used in cosmetic lotions. Isopropyl alcohol is used as a cleaning agent for keyboards, LCD displays and laptops. It is used as a wetting agent in the fountain solution used in lithographic printing and is often used as a solvent for french polishing shellac, which is used in cabinet making. It acts as a hydride source in the popular Meerwein-Ponndorf-Verley reduction. It is used in huge quantities as a preferred solvent in the manufacture of fine chemicals, drugs, drug intermediates and formulations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
MDL Number MFCD00011674
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Melting Point -90°C
Color Colorless
Density 0.785 g/mL
Boiling Point 82°C to 83°C
Flash Point 12°C (53°F)
Odor Alcohol-like
Packaging ChemSeal™ bottles
Linear Formula (CH3)2CHOH
Refractive Index 1.377
Quantity 250 mL
UN Number UN1219
Beilstein 635639
Merck Index 14,5208
Solubility Information Miscible with water,ethanol,acetone,chloroform,methanol,diethyl ether,n-octanol and benzene.
Formula Weight 60.1
Percent Purity ≥99.5%
Physical Form Liquid
Chemical Name or Material 2-Propanol, Anhydrous
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.