missing translation for 'onlineSavingsMsg'
Learn More

Solstice 2-Propanol

Product Code. 16625833
Change view
Click to view available options
Quantity:
7 L
18 L
200 L
Unit Size:
18 L
200 L
7 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16625833 7 L 7 L
15945470 18 L 18 L
15955470 200 L 200 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16625833 Supplier Solstice Supplier No. 33539S7LRC
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

2-Propanol, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99.8% (GC).

  • ACS general use solvents.
  • Value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis.
  • Suitable for less demanding applications such as routine isocratic HPLC.
  • Variations and decrease issues traceable to solvent contamination.
  • All solvents are tested to meet ACS general use specifications.

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
MDL Number MFCD00011674
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

pH Neutral
Melting Point -90°C
CAS Min % 100.00%
Color Colorless
CAS Max % 100.00%
Density 0.7855 g/cm3 (20°C)
Quantity 7 L
Flash Point 12°C
Packaging Returnable Stainless Steel Can
Physical Form Liquid
Product Line Puriss. p.a.
Linear Formula (CH3)2CHOH
Vapor Pressure 42 hPa (20°C)
Viscosity 2.66 mm2/s (25°C)
UN Number UN1219
Beilstein 635639
Concentration ≥99.80%
Synonym IPA,Isopropyl alcohol
Boiling Point 77°C to 83°C (1,013 hPa)
Percent Purity ≥99.80%
Grade ACS
Chemical Name or Material 2-Propanol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.