missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Propanethiol, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | 2-Propanethiol |
| CAS | 75-33-2 |
| Molecular Formula | C3 H8 S |
| Quantity | 5 g |
| Synonym | 2-Propanethiol (8CI','9CI','ACI), 1-Methylethanethiol, 2-Mercaptopropane, 2-Propylmercaptan, BHR 6986, i-Propanethiol, i-Propylthiol, Isopropanethiol, Isopropyl mercaptan, Isopropylthiol, NSC 87537 |
| InChI Formula | InChI=1S/C3H8S/c1-3(2)4/h3-4H,1-2H3 |
| SMILES | CC(C)S |
| IUPAC Name | propane-2-thiol |
| Molecular Weight (g/mol) | 76.16 |
| Formula Weight | 76.0347 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction